Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystathionase Antibody (OTI1E12), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB27048J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27048J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB27048J549 Supplier Novus Biologicals Supplier No. NBP270488JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cystathionase Monoclonal specifically detects Cystathionase in Human, Mouse, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cystathionase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E12
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cystathionase (cystathionine gamma-lyase), cystathionine gamma-lyase, cysteine desulfhydrase, EC 4.4.1, EC 4.4.1.1, gamma-cystathionase, homoserine deaminase, homoserine dehydratase, MGC9471
Gene Symbols CTH
Host Species Mouse
Immunogen Full length recombinant protein of human Cystathionase (NP_001893) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1491.0
Target Species Human, Mouse, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.