Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin A, Mouse anti-Human, Clone: CSTA/2882, Novus Biologicals™
SDP

Catalog No. NB010512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010512 20 μg
NB010511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010512 Supplier Novus Biologicals Supplier No. NBP27974720UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Cystatin A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CSTA/2882
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P01040, P01040
Gene Alias cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A
Gene Symbols CSTA
Host Species Mouse
Immunogen Recombinant fragment of human Cystatin A protein (around aa 1-98) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1475
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.