Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin A, Mouse anti-Human, Clone: CSTA/2882, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Cystatin A |
---|---|
Clone | CSTA/2882 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cystatin A | |
0.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
CSTA | |
IgG1 κ | |
Protein A or G purified |
CSTA/2882 | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01040, P01040 | |
1475 | |
Recombinant fragment of human Cystatin A protein (around aa 1-98) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title