Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin B/Stefin B Antibody (11H4), Novus Biologicals™
SDP

Catalog No. NB297695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297695 100 μL
NB297694 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297695 Supplier Novus Biologicals Supplier No. NBP326641100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cystatin B/Stefin B Monoclonal antibody specifically detects Cystatin B/Stefin B in Human samples. It is validated for ELISA, Immunocytochemistry

Specifications

Antigen Cystatin B/Stefin B
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 11H4
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Cystatin B/Stefin B [UniProt P04080]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1476
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.