Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin B/Stefin B Antibody (11H4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Cystatin B/Stefin B |
---|---|
Clone | 11H4 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Cystatin B/Stefin B Monoclonal antibody specifically detects Cystatin B/Stefin B in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Cystatin B/Stefin B | |
ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
A synthesized peptide derived from Human Cystatin B/Stefin B [UniProt P04080] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
11H4 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1476 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title