Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin C Antibody (Cyst19cc), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB004170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004170 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004170 Supplier Novus Biologicals Supplier No. NBP105766AF405

Mouse Monoclonal Antibody

Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Antigen Cystatin C
Applications ELISA
Classification Monoclonal
Clone Cyst19cc
Conjugate Alexa Fluor 405
Dilution ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Gene Symbols CST3
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Epitope Tags, Signal Transduction
Primary or Secondary Primary
Test Specificity Detects human Cystatin C, a member of the type 2 cystatin superfamily. Cystatin C is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsines), and pathogenic.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.