Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin C Antibody (OTI2C8), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270472B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270472B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270472B Supplier Novus Biologicals Supplier No. NBP270472B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cystatin C
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C8
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Gene Symbols CST3
Host Species Mouse
Immunogen Full length recombinant protein of human Cystatin C (NP_000090) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Epitope Tags, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1471.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.