Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin E/M/CST6 Antibody (037), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB077209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077209 Supplier Novus Biologicals Supplier No. NBP290741AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Cystatin E/M/CST6
Applications ELISA
Classification Monoclonal
Clone 37
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cystatin E/M/CST6 (NP_082899.1; Met1-Ala152).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1474
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.