Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin E/M/CST6 Antibody (MM0231-4H54), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB361174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361174 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361174 Supplier Novus Biologicals Supplier No. NBP212251MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Neutralization

Specifications

Antigen Cystatin E/M/CST6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Neutralization
Classification Monoclonal
Clone MM0231-4H54
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Host Species Mouse
Immunogen Human recombinant Cystatin E/M/CST6
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1474
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.