Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin E/M/CST6 Antibody (RM0217-6F49), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP212252IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212252IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212252IR Supplier Novus Biologicals Supplier No. NBP212252IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Cystatin E/M/CST6
Applications Western Blot
Classification Monoclonal
Clone RM0217-6F49
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Gene Symbols CST6
Host Species Rat
Immunogen Mouse recombinant Cystatin E/M/CST6
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1474
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.