Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin F Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB321265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321265 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321265 Supplier Novus Biologicals Supplier No. NBP298244APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cystatin F Polyclonal antibody specifically detects Cystatin F in Mouse samples. It is validated for ELISA

Specifications

Antigen Cystatin F
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Cystatin F (Uniprot#: O89098; Met1-Gln144)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 8530
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.