Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin S Antibody (OTI2H10), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP270481C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270481C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270481C Supplier Novus Biologicals Supplier No. NBP270481C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cystatin S Monoclonal specifically detects Cystatin S in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cystatin S
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2H10
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1
Gene Symbols CST4
Host Species Mouse
Immunogen Full length recombinant protein of human Cystatin S (NP_001890) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1472.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.