Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin SN Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB089670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089670 Supplier Novus Biologicals Supplier No. NBP299826F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cystatin SN Polyclonal specifically detects Cystatin SN in Human samples. It is validated for ELISA.

Specifications

Antigen Cystatin SN
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cystatin SN (P01037-1; Met 1-Ser 141).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 1469
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.