Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SN Antibody (MM0234-5N51) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Cystatin SN |
---|---|
Clone | MM0234-5N51 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot.Specifications
Cystatin SN | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P01037 | |
1469 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0234-5N51 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
CST1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title