Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin SN Rabbit anti-Human, Clone: 111, Novus Biologicals™
SDP

Catalog No. NB021304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021304 100 μg
NB021305 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021304 Supplier Novus Biologicals Supplier No. NBP289919100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Cystatin SN
Applications ELISA
Classification Monoclonal
Clone 111
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family
Host Species Rabbit
Immunogen Recombinant Human Cystatin SN protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1469
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.