Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Dioxygenase Type 1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB184844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184844 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184844 Supplier Novus Biologicals Supplier No. NBP282081C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cysteine Dioxygenase Type 1 Polyclonal antibody specifically detects Cysteine Dioxygenase Type 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Cysteine Dioxygenase Type 1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CDO, CDO-I, cysteine dioxygenase type 1, Cysteine dioxygenase type I, cysteine dioxygenase, type I, EC 1.13.11.20
Host Species Rabbit
Immunogen Cysteine Dioxygenase Type 1 antibody was raised against a 19 amino acid peptide near the amino terminus of human Cysteine Dioxygenase Type 1. The immunogen is located within amino acids 30 - 80 of Cysteine Dioxygenase Type 1. Amino Acid Squence: DPTEWAMYAKFDQYRYTRN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1036
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.