Learn More
Epredia™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunofluorescence and immunoprecipitation experiments.
$260.00
Specifications
Antigen | Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-3 |
---|---|
Clone | L12B4 |
Applications | Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
MS1248P0
|
Epredia™
MS1248P0 |
200μg/mL | 100 μL |
Each for $260.00
|
|
||||
MS1248P1
|
Epredia™
MS1248P1 |
200μg/mL | 500 μL | N/A | N/A | N/A | |||
MS1248P
|
Epredia™
MS1248P |
200μg/mL | 1 mL | N/A | N/A | N/A | |||
MS1248P1ABX
|
Epredia™
MS1248P1ABX |
1mg/mL | 100 μL | N/A | N/A | N/A | |||
MS1248PABX
|
Epredia™
MS1248PABX |
1mg/mL | 200 μL | N/A | N/A | N/A | |||
Description
CFTR (cystic fibrosis transmembrane conductance regulator) is composed of two membrane-spanning domains (MSD), two nucleotide-binding domains (NBD), and an R domain. It is structurally similar to multidrug resistance (Mdr1) protein and both are members of the superfamily of ATP-binding cassette (ABC) transporters, also known as traffic ATPases, which are implicated in the movement of various substrates. The CFTR protein is a small conductance adenosine 3',5'-cyclic monophosphate (cAMP)-activated chloride ion channel found in the apical membranes of epithelia within the pancreas, airway, intestine, bile duct, sweat gland, and male genital ducts. CFTR is a valuable marker of human pancreatic duct cell development and differentiation.Host Species: Mouse
Clone: L12B4
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 386-412
Immunogen: Recombinant protein encoding NBF1 domain of human CFTR
Molecular Weight: 165-170 kDa
Positive Control: Pancreas
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not verified)
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-3 | |
Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
IgG1 |
L12B4 | |
Monoclonal | |
Concentrated | |
RUO | |
Recombinant protein encoding NBF1 domain of human CFTR | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.