Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYTIP Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB101976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101976 Supplier Novus Biologicals Supplier No. NBP297434JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYTIP Polyclonal specifically detects CYTIP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CYTIP
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias B3-1, CASPCytohesin-associated scaffolding protein, Cbp HE, CYBRCytohesin-binding protein HE, CYTHIP, cytohesin 1 interacting protein, Cytohesin binder and regulator, cytohesin-1 interacting protein, cytohesin-interacting protein, HEPleckstrin homology Sec7 and coiled-coil domains-binding protein, pleckstrin homology, Sec7 and coiled/coil domains, binding protein, pleckstrin homology, Sec7 and coiled-coil domains, binding protein, PSCDBPcytohesin binding protein HE
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYTIP fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9595
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.