Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cytochalasin D
GSA_VAGREENER_CHOICE

Catalog No. p-4783387
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Disrupts actin filament function

Potent disruptor of actin filament function. Alters tight junction permeability. Unlike cytochalasin B (Cat. No. 5474), does not inhibit monosaccharide transport across the plasma membrane.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 22144-77-0
Molecular Formula C30H37NO6

Specifications

Quantity 1 mg
Formula Weight 507.63
Purity >95%
Molecular Weight (g/mol) 507.627
Chemical Name or Material Cytochalasin D
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.