Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome b245 alpha Antibody (CS9), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP140974UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140974UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140974UV Supplier Novus Biologicals Supplier No. NBP140974UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome b245 alpha Monoclonal specifically detects Cytochrome b245 alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Cytochrome b245 alpha
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CS9
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Accession No. P13498
Gene Alias cytochrome b light chain, Cytochrome b(558) alpha chain, cytochrome b(558) alpha-subunit, cytochrome b, alpha polypeptide, cytochrome b-245 light chain, cytochrome b-245, alpha polypeptide, Cytochrome b558 subunit alpha, flavocytochrome b-558 alpha polypeptide, Neutrophil cytochrome b 22 kDa polypeptide, p22 phagocyte B-cytochrome, p22phox, p22-phox, Superoxide-generating NADPH oxidase light chain subunit
Gene Symbols CYBA
Host Species Mouse
Immunogen Cytochrome B245 solubilised in octyl-beta-gluycopyranoside
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1535
Test Specificity This antibody recognizes the light chain of cytochrome B245, also known as p22phox, a subunit of the nicotinamide adenine dinucleotide phosphate-oxidase (NADPH oxidase). NADPH oxidase is a membrane-bound enzyme complex in neutrophils that produces the superoxide anion (O2-) by transferring single electrons from NADPH inside the cell across the membrane and coupling them to molecular oxygen. Superoxide is used for the destruction of pathogens. Individuals with defects in NADPH oxidase suffer from chronic granulomatous disease, characterized by severe recurrent infections.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.