Learn More
Description
Specifications
Specifications
| Antigen | Cytochrome b245 alpha |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | CS9 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen |
| Gene Accession No. | P13498 |
| Gene Alias | cytochrome b light chain, Cytochrome b(558) alpha chain, cytochrome b(558) alpha-subunit, cytochrome b, alpha polypeptide, cytochrome b-245 light chain, cytochrome b-245, alpha polypeptide, Cytochrome b558 subunit alpha, flavocytochrome b-558 alpha polypeptide, Neutrophil cytochrome b 22 kDa polypeptide, p22 phagocyte B-cytochrome, p22phox, p22-phox, Superoxide-generating NADPH oxidase light chain subunit |
| Gene Symbols | CYBA |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
