Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome b5 Outer Mitochondrial Membrane Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB042956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042956 Supplier Novus Biologicals Supplier No. NBP297403AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytochrome b5 Outer Mitochondrial Membrane Polyclonal specifically detects Cytochrome b5 Outer Mitochondrial Membrane in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytochrome b5 Outer Mitochondrial Membrane
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CYB5M, CYB5-M, CYPB5M, Cytochrome b5 outer mitochondrial membrane isoform, cytochrome b5 type B, cytochrome b5 type B (outer mitochondrial membrane), DKFZp686M0619, OMB5, outer mitochondrial membrane cytochrome b5, type 2 cyt-b5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Cytochrome b5 Outer Mitochondrial Membrane fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.