Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Antibody (6H2.B4), Novus Biologicals™
SDP

Catalog No. NBP224872 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224872 0.1 mg
NBP224872SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224872 Supplier Novus Biologicals Supplier No. NBP224872
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat samples. It is validated for Block/Neutralize, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Cytochrome c
Applications Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 6H2.B4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry reported in scientific literature (PMID 28194435), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16540654), Immunoprecipitation 2 ug/1X10^6 cells, Block/Neutralize
Gene Accession No. P99999
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbols CYCS
Host Species Mouse
Immunogen Rat cytochrome c was used as immunogen to generate 6H2.B4 hybridoma clone (Goshorn et al, 1991). The antibody recognizes an epitope around residue 62 of rat Cytochrome c, based on competitive ELISA results (Goshorn et al. 1991).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54205
Test Specificity This antibody is reactive to Human, Mouse and Rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.