Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Antibody (6H2.B4), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$206.50 - $499.50

Specifications

Antigen Cytochrome c
Clone 6H2.B4
Concentration 0.5 mg/mL
Dilution Immunohistochemistry reported in scientific literature (PMID 28194435), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16540654), Immunoprecipitation 2 ug/1X10^6 cells, Block/Neutralize
Applications Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224872SS
SDP
View Documents
Novus Biologicals
NBP224872SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224872
SDP
View Documents
Novus Biologicals
NBP224872
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize.
Specifications

Specifications

Cytochrome c
0.5 mg/mL
Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
P99999
54205
Rat cytochrome c was used as immunogen to generate 6H2.B4 hybridoma clone (Goshorn et al, 1991). The antibody recognizes an epitope around residue 62 of rat Cytochrome c, based on competitive ELISA results (Goshorn et al. 1991).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
6H2.B4
Immunohistochemistry reported in scientific literature (PMID 28194435), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16540654), Immunoprecipitation 2 ug/1X10^6 cells, Block/Neutralize
Monoclonal
Purified
RUO
Human, Mouse, Rat
CYCHCS, cytochrome c, cytochrome c, somatic, THC4
CYCS
IgG1 κ
Protein G purified
This antibody is reactive to Human, Mouse and Rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.