Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Antibody (7H8.2C12 + CYCS/1010), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB007154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007154 Supplier Novus Biologicals Supplier No. NBP247694APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Cytochrome c
Applications Flow Cytometry
Classification Monoclonal
Clone 7H8.2C12 + CYCS/1010
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbols CYCS
Host Species Mouse
Immunogen Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c (7H8.2C12); Recombinant full-length human CYCS protein (CYCS/1010) (Uniprot: P99999)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Cytochrome C is a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of cytochrome c, thereby blocking the apoptotic process. Overexpression of Bax has been shown to induce the release of cytochrome c and to induce cell death. The release of cytochrome c from the mitochondria is thought to trigger an apoptotic cascade, whereby Apaf-1 binds to Apaf-3 (caspase-9) in a cytochrome c-dependent manner, leading to caspase-9 cleavage of caspase-3. This monoclonal antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached).
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.