Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Antibody (7H8.2C12), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10056503V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056503V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056503V Supplier Novus Biologicals Supplier No. NB10056503V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome c Monoclonal specifically detects Cytochrome c in Drosophila, Human, Mouse, Rabbit, Rat samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Cytochrome c
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7H8.2C12
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. P99999
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbols CYCS
Host Species Mouse
Immunogen Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54205
Test Specificity This antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached).
Target Species Drosophila, Human, Rat, Mouse, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.