Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Antibody (rCYCS/1010), FITC, Novus Biologicals™
SDP

Catalog No. NB385740 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385740 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385740 Supplier Novus Biologicals Supplier No. NBP308588F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome c Monoclonal antibody specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytochrome c
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rCYCS/1010
Conjugate FITC
Formulation PBS
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Host Species Mouse
Immunogen Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c (Uniprot: P99999)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54205
Target Species Human, Mouse, Rat, Amphibian, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.