Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C Oxidase subunit 6c Antibody, Novus Biologicals™
SDP

Catalog No. NB397067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397067 25 μL
NBP237857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397067 Supplier Novus Biologicals Supplier No. NBP23785725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytochrome C Oxidase subunit 6c Polyclonal specifically detects Cytochrome C Oxidase subunit 6c in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytochrome C Oxidase subunit 6c
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P09669
Gene Alias Cytochrome c oxidase polypeptide VIc, cytochrome c oxidase subunit 6C, cytochrome c oxidase subunit VIc, cytochrome c oxidase subunit VIc preprotein
Gene Symbols COX6C
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: EPRKKAYADFYRNYDVMKDFEDMRKTGT
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1345
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.