Learn More
Cytochrome C Oxidase subunit 6c Antibody (OTI4B11), Alexa Fluor™ 700, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytochrome C Oxidase subunit 6c |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI4B11 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Cytochrome c oxidase polypeptide VIc, cytochrome c oxidase subunit 6C, cytochrome c oxidase subunit VIc, cytochrome c oxidase subunit VIc preprotein |
| Gene Symbols | COX6C |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.