Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 17A1 Antibody (OTI3F12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27042A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27042A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27042A94 Supplier Novus Biologicals Supplier No. NBP270542AF594

Mouse Monoclonal Antibody

Cytochrome P450 17A1 Monoclonal specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cytochrome P450 17A1
Applications Western Blot
Classification Monoclonal
Clone OTI3F12
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH
Gene Symbols CYP17A1
Host Species Mouse
Immunogen Full length recombinant protein of human Cytochrome P450 17A1 (NP_000093) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1586.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.