Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 17A1 Antibody (OTI3F12), PerCP, Novus Biologicals™
SDP

Catalog No. NB289527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289527 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289527 Supplier Novus Biologicals Supplier No. NBP270542PCP

Mouse Monoclonal Antibody

Cytochrome P450 17A1 Monoclonal antibody specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Western Blot

Specifications

Antigen Cytochrome P450 17A1
Applications Western Blot
Classification Monoclonal
Clone OTI3F12
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH
Host Species Mouse
Immunogen Full length human recombinant protein of human CYP17A1(NP_000093) peoduced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1586
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.