Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 17A1 Rabbit anti-Human, Mouse, Clone: 5D2H1, Novus Biologicals™
SDP

Catalog No. NB168174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168174 20 μg
NB168173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168174 Supplier Novus Biologicals Supplier No. NBP31654120UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytochrome P450 17A1 Monoclonal antibody specifically detects Cytochrome P450 17A1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Cytochrome P450 17A1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 5D2H1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 101-220 of human Cytochrome P450 17A1 (P05093).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1586
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.