Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 1A1/1A2 Antibody (MC1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB379047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379047 Supplier Novus Biologicals Supplier No. NBP308705AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 1A1/1A2 Monoclonal antibody specifically detects Cytochrome P450 1A1/1A2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Cytochrome P450 1A1/1A2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MC1
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias AHH, AHRR, aryl hydrocarbon hydroxylase, CP11, CYP1, CYPIA1, cytochrome P1-450, dioxin-inducible, cytochrome P450 1A1, Cytochrome P450 form 6, cytochrome P450, family 1, subfamily A, polypeptide 1, Cytochrome P450-C, Cytochrome P450-P1, EC 1.14.14.1, flavoprotein-linked monooxygenase, P1-450, P450-C, P450DX, subfamily I (aromatic compound-inducible), polypeptide 1, xenobiotic monooxygenase
Host Species Mouse
Immunogen 3-methylcholanthrene induced rat cytochrome P450 protein.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1543
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.