Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 1A2 Antibody (D15), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB6001490AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001490AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001490AF Supplier Novus Biologicals Supplier No. NB6001490AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 1A2 Monoclonal specifically detects Cytochrome P450 1A2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Cytochrome P450 1A2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone D15
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase
Gene Symbols CYP1A2
Host Species Mouse
Immunogen MC1a (Preparation C31B4, rat liver cytochrome P4501A2)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1544
Test Specificity D15 cross reacts with CYP1A1 & CYP1A2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.