Learn More
Cytochrome P450 1A2 Mouse anti-Human, Rat, Clone: 3B8C1, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytochrome P450 1A2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 3B8C1 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1 - 5 ug/ml |
| Formulation | PBS |
| Gene Alias | aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase |
| Host Species | Mouse |
| Immunogen | Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with rat cytochrome p450 proteins and mouse myeloma Ag8563 cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.