Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 26B1 Antibody (T5P3G2*A3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2019A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2019A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2019A30 Supplier Novus Biologicals Supplier No. NBP250195AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 26B1 Monoclonal specifically detects Cytochrome P450 26B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cytochrome P450 26B1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T5P3G2*A3
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome
Gene Symbols CYP26B1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 56603.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.