Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2A6, 2A7, 2A13 Antibody (F16 P2 D8), HRP, Novus Biologicals™
SDP

Catalog No. NBP250184H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250184H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250184H Supplier Novus Biologicals Supplier No. NBP250184H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 2A6, 2A7, 2A13 Monoclonal specifically detects Cytochrome P450 2A6, 2A7, 2A13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Cytochrome P450 2A6, 2A7, 2A13
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone F16 P2 D8
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with No Preservative
Host Species Mouse
Immunogen ovalbumin-conjugated synthetic peptide RNYTMSFLPR. CYP2A6, CYP2A7and CYP2A13 have the same c-terminal sequence. (C-terminal sequence identical in human and rhesus macaque)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.