Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2A6 Antibody (OTI1D2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270524APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270524APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270524APC Supplier Novus Biologicals Supplier No. NBP270524APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 2A6 Monoclonal specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen Cytochrome P450 2A6
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1D2
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase
Gene Symbols CYP2A6
Host Species Mouse
Immunogen Full length recombinant protein of human Cytochrome P450 2A6 (NP_000753) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1548.0
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.