Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2B1, 2B2 Antibody (b/e3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP250201V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250201V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250201V Supplier Novus Biologicals Supplier No. NBP250201V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 2B1, 2B2 Monoclonal specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Cytochrome P450 2B1, 2B2
Applications Western Blot, ELISA
Classification Monoclonal
Clone b/e3
Conjugate DyLight 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Rat liver cytochromes p450 2B1 and 2B2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Test Specificity Clone b/e3 inhibits PROD activity in liver microsomes of PB-treated rats and inhibits ECOD activity in liver microsomes of PB-treated but not 3MC-treated rats. The CYP2B1 antibody recognizes the major barbiturate-inducible P450s in mouse and rat.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.