Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2B1, 2B2 Antibody (b/e4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309780 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309780 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309780 Supplier Novus Biologicals Supplier No. NBP250202MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 2B1, 2B2 Monoclonal antibody specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, ELISA

Specifications

Antigen Cytochrome P450 2B1, 2B2
Applications Western Blot, ELISA
Classification Monoclonal
Clone b/e4
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Rat liver cytochromes P4502B1 and 2B2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.