Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2B6 Antibody (OTI3D5), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270526R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270526R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270526R Supplier Novus Biologicals Supplier No. NBP270526R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 2B6 Monoclonal specifically detects Cytochrome P450 2B6 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Cytochrome P450 2B6
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3D5
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6cytochrome P450, subfamily IIB (phenobarbital-inducible), cytochrome P450 2B6, Cytochrome P450 IIB1, cytochrome P450, family 2, subfamily B, cytochrome P450, family 2, subfamily B, polypeptide 6, cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6, EC 1.14.14.1, IIB1, P450
Gene Symbols CYP2B6
Host Species Mouse
Immunogen Full length recombinant protein of human Cytochrome P450 2B6 (NP_000758) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1555.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.