Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2C9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $617.01
Specifications
| Antigen | Cytochrome P450 2C19 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cytochrome P450 2C9 Polyclonal specifically detects Cytochrome P450 2C9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytochrome P450 2C19 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| (R)-limonene 6-monooxygenase, (S)-limonene 6-monooxygenase, (S)-limonene 7-monooxygenase, CPCJ, CYP2C, CYPIIC17, CYPIIC19, cytochrome P450 2C19, cytochrome P-450 II C, cytochrome P450, family 2, subfamily C, polypeptide 19, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19, Cytochrome P450-11A, Cytochrome P450-254C, EC 1.14.13.48, EC 1.14.13.49, EC 1.14.13.80, EC 1.14.14.1, flavoprotein-linked monooxygenase, mephenytoin 4'-hydroxylase, Mephenytoin 4-hydroxylase, microsomal monooxygenase, P450C2C, P450IIC19, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
| CYP2C19 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1557 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KEHQESMDINNPRDFIDCFLIKMEKEKQNQQSEFTIENLVITAADLLGAGTETTST | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title