Learn More
Cytochrome P450 2C9 Antibody (OTI1D7), Janelia Fluor™ 549, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cytochrome P450 2C9 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI1D7 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | (R)-limonene 6-monooxygenase, (S)-limonene 6-monooxygenase, (S)-limonene 7-monooxygenase, CPC9, CYP2C, CYP2C10, CYPIIC9, cytochrome P450 2C9, Cytochrome P450 MP-4, Cytochrome P450 MP-8, Cytochrome P450 PB-1, cytochrome P-450 S-mephenytoin 4-hydroxylase, cytochrome P450, family 2, subfamily C, polypeptide 9, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 9, Cytochrome P-450MP, EC 1.14.13.48, EC 1.14.13.49, EC 1.14.13.80, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC149605, MGC88320, microsomal monooxygenase, P450IIC9, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase |
| Gene Symbols | CYP2C9 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.