Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2D10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229340 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229340 Supplier Novus Biologicals Supplier No. NBP229340
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Cytochrome P450 2D10 Polyclonal specifically detects Cytochrome P450 2D10 in Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Cytochrome P450 2D10
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_775426.1
Gene Alias 1.14.14.1, Cyp2d10, Cyp2d-10, Cyp2d5, CYPIID10, Cytochrome P450-CMF1B, Cytochrome P450-DB5, Debrisoquine 4-hydroxylase
Gene Symbols Cyp2d5
Host Species Goat
Immunogen Peptide with sequence CDAFTAQNGRSINPK corresponding to internal region according to NP_775426.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286963
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.