Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2D6 Rabbit anti-Human, Mouse, Rat, Clone: 1X3F0, Novus Biologicals™
SDP

Catalog No. NB168311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168311 100 μg
NB168312 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168311 Supplier Novus Biologicals Supplier No. NBP316610100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytochrome P450 2D6 Monoclonal antibody specifically detects Cytochrome P450 2D6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Cytochrome P450 2D6
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1X3F0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CPD6P450DB1, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, cytochrome P450 2D6, cytochrome P450, family 2, subfamily D, polypeptide 6, cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2, cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2, cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), Cytochrome P450-DB1, Debrisoquine 4-hydroxylase, EC 1.14.14.1, flavoprotein-linked monooxygenase, -metabolizing)-like 1, MGC120389, MGC120390, microsomal monooxygenase, P450C2D, P450-DB1, polypeptide 6, polypeptide 7 pseudogene 2, polypeptide 8 pseudogene 2, xenobiotic monooxygenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 341-497 of human Cytochrome P450 2D6 (P10635).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1565
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.