Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome p450 2J2 Antibody (OTI5C9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP270533 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270533 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270533 Supplier Novus Biologicals Supplier No. NBP270533
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome p450 2J2 Monoclonal specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cytochrome p450 2J2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5C9
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:500
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase
Gene Symbols CYP2J2
Host Species Mouse
Immunogen Full length human recombinant protein of human CYP2J2(NP_000766) produced in HEK293T cell.
Molecular Weight of Antigen 57.4 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1573
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.