Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 307a1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178150 0.1 mg
NB018754 0.025 mg
NBP17815005 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP178150 Supplier Novus Biologicals Supplier No. NBP178150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytochrome P450 307a1 Polyclonal specifically detects Cytochrome P450 307a1 in Drosophila samples. It is validated for Western Blot, ELISA, Dot Blot.

Specifications

Antigen Cytochrome P450 307a1
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration 0.60 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Dot Blot 1:10000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Accession No. AAQ05973
Gene Alias 307a1, CG10594, Cyp307a1, CYP307A1-Dm, Dmel\CG10594, Dmspo, Spo, spo spook
Host Species Rabbit
Immunogen Synthetic peptide taken form the Spook protein form within the C-terminal region (amino acid 500-543), the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Spook peptide sequence was conserved in rat, mouse and human protein and also present in other species.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38631
Test Specificity NBP1-78150 recognizes Drosophila Cytochrome P450 307a1 .
Target Species Drosophila
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.