Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 307a1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
Antigen | Cytochrome P450 307a1 |
---|---|
Concentration | 0.60 mg/mL |
Dilution | Western Blot 1:500, ELISA 1:10000, Dot Blot 1:10000 |
Applications | Dot Blot, Western Blot, ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018754
![]() |
Novus Biologicals
NBP1781500.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP17815005
![]() |
Novus Biologicals
NBP1781500.05MG |
0.05 mg |
Each for $402.00
|
|
|||||
NBP178150
![]() |
Novus Biologicals
NBP178150 |
0.1 mg |
Each for $670.50
|
|
|||||
Description
Cytochrome P450 307a1 Polyclonal specifically detects Cytochrome P450 307a1 in Drosophila samples. It is validated for Western Blot, ELISA, Dot Blot.Specifications
Cytochrome P450 307a1 | |
Western Blot 1:500, ELISA 1:10000, Dot Blot 1:10000 | |
Polyclonal | |
Rabbit | |
AAQ05973 | |
38631 | |
IgG |
0.60 mg/mL | |
Dot Blot, Western Blot, ELISA | |
Unconjugated | |
Drosophila | |
307a1, CG10594, Cyp307a1, CYP307A1-Dm, Dmel\CG10594, Dmspo, Spo, spo spook | |
Synthetic peptide taken form the Spook protein form within the C-terminal region (amino acid 500-543), the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Spook peptide sequence was conserved in rat, mouse and human protein and also present in other species. | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title