Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 3A4/3A5 Antibody (F24P2B10), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250208FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250208FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250208FR Supplier Novus Biologicals Supplier No. NBP250208FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 3A4/3A5 Monoclonal specifically detects Cytochrome P450 3A4/3A5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Cytochrome P450 3A4, 3A5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone F24P2B10
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide ESRDGTVSGA
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.