Learn More
Cytochrome P450 3A4/3A5 Antibody (F24P2B10), Janelia Fluor™ 646, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CytochromeP4503A4,3A5 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | F24P2B10 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Host Species | Mouse |
| Immunogen | Ovalbumin-conjugated synthetic peptide ESRDGTVSGA |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.